Iodomethane 2 M in tert-butyl methyl ether, AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl iodide

378411000 378418000
TS378411000EA 120 USD
TS378411000 TS378418000
Iodomethane 2 M in tert-butyl methyl ether, AcroSeal®
Iodomethane
Formula: CH₃I
Flash Pt: –18 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001073
CAS Number: 74-88-4
EINECS: 200-819-5
UN: 1992
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Solution
Stabilizer Silver
Strength 1.90 to 2.20 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR