Tin(IV) chloride 1 mol/l in dichloromethane, AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Stannic chloride, Tin tetrachloride

378201000 378208000
TS378201000EA 68 USD
TS378201000 TS378208000
Tin(IV) chloride 1 mol/l in dichloromethane, AcroSeal®
Tin (IV) chloride
Formula: Cl₄Sn
Storage Temperature: Ambient
MDL Number: MFCD00011242
CAS Number: 7646-78-8
EINECS: 231-588-9
UN: 2922

Order Now

Specification Test Results

Appearance Clear colorless Solution
Titration Argentometric 0.95 to 1.10 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR