Lithium tri-tert-butoxyaluminum hydride 1.1 M in THF, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lithium tri-tert-butoxyaluminohydride

377588000
TS37758-1000EA 151 USD
TS37758-1000 TS37758-8000
Lithium tri-tert-butoxyaluminum hydride 1.1 M in THF, AcroSeal™
Lithium tri-tert-butoxyaluminum hydride
Formula: C₁₂H₂₈AlLiO₃
Density: 0.9 g/cm³
Flash Pt: –17 °C
Storage Temperature: Ambient
MDL Number: MFCD00011532
CAS Number: 17476-04-9
UN: 3399
ADR: 4.3,I

Order Now

Specification Test Results

Appearance Clear to hazy colorless Solution (May develop some turbidity or precipitate)
Assay 28 to 32 % (LTTBA)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR