Potassium thiocyanate 98%, pure

Supplier: Thermo Scientific Chemicals
Warning

AC37743-0025 AC37743-1000 AC37743-5000
TS37743-0025EA 169 USD
TS37743-0025 TS37743-1000 TS37743-5000
Potassium thiocyanate 98%, pure
Potassium thiocyanate
Formula: KSCN
Melting Pt: 170…179 °C
Density: 1.886 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011413
CAS Number: 333-20-0
EINECS: 206-370-1
Merck Index: 12,07866

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR