N-(tert-Butoxycarbonyl)-trans-4-hydroxy-L-prolinemethyl ester 97%

Supplier: Thermo Scientific Chemicals
Warning

37625-0050
TS37625-0050EA 97 USD
TS37625-0050
N-(tert-Butoxycarbonyl)-trans-4-hydroxy-L-prolinemethyl ester 97%
N-(tert-Butoxycarbonyl)-trans-4-hydroxy-L-prolinemethyl ester
Formula: C₁₁H₁₉NO₅
Melting Pt: 91…96 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00076981
CAS Number: 74844-91-0

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
GC ≥96.0 %
Specific optical rotation -63° to -67° (20°C, 589 nm) (c=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR