4-Hydroxy-6-methylcoumarin 98+%

Supplier: Thermo Scientific Chemicals
Warning

37287-0010 37287-0050
TS37287-0010EA 29 USD
TS37287-0010 TS37287-0050
4-Hydroxy-6-methylcoumarin 98+%
4-Hydroxy-6-methylcoumarin
Formula: C₁₀H₈O₃
Melting Pt: 261…264 °C
Storage Temperature: Ambient
MDL Number: MFCD00085738
CAS Number: 13252-83-0

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 258 to 262 °C
Titration with TBAH ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR