Hydrogen chloride 1 M in acetic acid, AcroSeal®

Supplier: Thermo Scientific Chemicals
Danger

370271000
TS370271000EA 41 USD
TS370271000
Hydrogen chloride 1 M in acetic acid, AcroSeal®
Hydrogen chloride
Formula: HCl
Density: 1.092 g/cm³
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 2920
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless Solution
Strength 1 M Exact strength on the certificate of analysis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR