Strontium chloride, anhydrous 99.99% (trace metals basis)

Supplier: Thermo Scientific Chemicals
Danger

AC36974-0050 AC36974-0250
TS36974-0050EA 106 USD
TS36974-0050 TS36974-0250
Strontium chloride, anhydrous 99.99% (trace metals basis)
Strontium chloride
Formula: Cl₂Sr
MW: 158.53
Boiling Pt: 1250
Melting Pt: 868
Density: 1.96
MDL Number: MFCD00011249
CAS Number: 10476-85-4
EINECS: 233-971-6
Merck Index: 12,09000

Order Now

Specification Test Results

Appearance White Crystalline powder
Assay 54.4 to 55.9 % (Sr)
Total trace metal impurities ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR