4-Hydroxyphenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(4-Hydroxyphenyl)metaboric acid, Dihydroxy(4-hydroxyphenyl)borane, 4-Hydroxybenzeneboronic acid
36942-0010
36942-0050
TS36942-0010EA
55
USD
TS36942-0010
TS36942-0050
4-Hydroxyphenylboronic acid 97%
4-Hydroxyphenylboronic acid
Formula:
C₆H₇BO₃ MW: 137.93 g/mol Melting Pt: 285 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD01074628 CAS Number: 71597-85-8 |
Specification Test Results
Appearance | Off-white to light brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...