tert-Butylchlorodimethylsilane 1.0 M in dichloromethane, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: tert-Butyldimethylsilyl Chloride, Chloro-tert-butyldimethylsilane, t-Butyldimethylchlorosilane, tert-Butyldimethylchlorosilane, tert-Butyldimethylsilyl chloride, TBDMS chloride

AC36910-1000
TS36910-1000EA 124 USD
TS36910-1000
tert-Butylchlorodimethylsilane 1.0 M in dichloromethane, AcroSeal™
tert-Butylchlorodimethylsilane
Formula: C₆H₁₅ClSi
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00000501
CAS Number: 18162-48-6
EINECS: 242-042-4
UN: 2920
ADR: 8,I

Order Now

Specification Test Results

Appearance Colorless Solution
Infrared spectrum Conforms
Titration Argentometric 0.95 to 1.10 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR