Hydrogen chloride 1 N diethyl ether, AcroSeal™, pure

Supplier: Thermo Scientific Chemicals
Danger

36846-1000 368468000
TS36846-1000EA 64 USD
TS36846-1000 TS368468000
Hydrogen chloride 1 N diethyl ether, AcroSeal™, pure
Hydrogen chloride
Formula: HCl
MW: 36.46
Density: 0.74 g/cm³
Flash Pt: –40 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 2924
ADR: 3 I

Order Now

Specification Test Results

Appearance Clear colorless Solution
Strength 0.95 to 1.10 N


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR