1-(2-Methoxyethyl)piperazine 98%

Supplier: Thermo Scientific Chemicals
Danger

36711-0010
TS36711-0010EA 41 USD
TS36711-0010 TS36711-0050
1-(2-Methoxyethyl)piperazine 98%
1-(2-Methoxyethyl)piperazine
Formula: C₇H₁₆N₂O
Flash Pt: 87 °C
MDL Number: MFCD00191214
CAS Number: 13484-40-7
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4720 to 1.4750 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR