Triphenylphosphine 1,2 - 1,5 mmol/g resin, cross-linked with 1% DVB 200-400 mesh

Supplier: Thermo Scientific Chemicals
Warning

36684-0050
TS36684-0050EA 93 USD
TS36684-0050
Triphenylphosphine 1,2 - 1,5 mmol/g resin, cross-linked with 1% DVB 200-400 mesh
Triphenylphosphine
Formula: C₁₈H₁₅P
Storage Temperature: Refrigerator
MDL Number: MFCD00003043
CAS Number: 603-35-0

Order Now

Specification Test Results

Appearance White to yellow Beads
Particle size 200 to 400 mesh
Crosslinking 1 % DVB
Loading 1 to 1.5 mmol/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR