2-(3-Trifluoromethylphenyl)ethylamine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
3-(Trifluoromethyl)phenethylamine
36552-0010
36552-0050
TS36552-0010EA
24
USD
TS36552-0010
TS36552-0050
2-(3-Trifluoromethylphenyl)ethylamine 98%
2-(3-Trifluoromethylphenyl)ethylamine
Formula:
C₉H₁₀F₃N Storage Temperature: Ambient |
MDL Number:
MFCD00040756 CAS Number: 52516-30-0 UN: 2735 ADR: 8,III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4635 to 1.4655 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...