n-Heptane ∼ 99% (n-heptane) for HPLC

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: high purity solvents

364970010 364970025
TS364970010EA 123 USD
TS364970010 TS364970025
n-Heptane ∼ 99% (n-heptane) for HPLC
n-Heptane
Formula: H₃C(CH₂)₅CH₃
Boiling Pt: 98.4 °C (1013 hPa)
Melting Pt: –91 °C
Density: 0.683 g/cm³
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.01 %
GC ≥99 %
UV at 230 nm A: ≤0.02
Residue after evaporation ≤5 ppm
Acidity ≤0.00015 meq/g
Filtered to 0.2 micrometer Filtered


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR