N-Methyl-2-pyrrolidone (NMP), extra dry over Molecular Sieve 99.5%, AcroSeal™

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, N-Methylpyrrolidinone, N-Methyl-2-pyrrolidinone, N-Methyl-alpha-pyrrolidone, M-Pyrol, N-methylpyrrolidone, N-Methyl-gamma-butyrolactam, 1-Methyl-2-pyrrolidone, NMP, N-Methylpyrrolidone, N-Methyl-2-pyrrolidone, Methyl Pyrrolidone

36438-1000 36438-5000 364380010 364380025
TS36438-1000EA 104 USD
TS36438-1000 TS36438-5000 TS364380010 TS364380025
N-Methyl-2-pyrrolidone (NMP), extra dry over Molecular Sieve 99.5%, AcroSeal™
N-Methyl-2-pyrrolidone (NMP)
Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 202
Melting Pt: -24
Density: 1.03 g/cm³
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid (May darken on storage)
Infrared spectrum Conforms
Water ≤0.005 % (Coulometric)
GC ≥99.5 %
Color scale ≤50 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR