Scandium (III) trifluoromethanesulfonate 95%

Supplier: Thermo Scientific Chemicals
Danger

AC36389-0010 AC36389-0050
TS36389-0010EA 70 USD
TS36389-0010 TS36389-0050
Scandium (III) trifluoromethanesulfonate 95%
Scandium (III) trifluoromethanesulphonate
Formula: C₃F₉O₉S₃Sc
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00192433
CAS Number: 144026-79-9
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Elemental Analysis ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR