Tris(hydroxymethyl)phosphine 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
36384-0010
36384-0250
TS36384-0010EA
79
USD
TS36384-0010
TS36384-0250
Tris(hydroxymethyl)phosphine 95%
Tris(hydroxymethyl)phosphine
Formula:
C₃H₉O₃P Boiling Pt: 111…113 °C (2.5 mmHg) Melting Pt: 48…56 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00055382 CAS Number: 2767-80-8 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | White to light yellow Powder or crystals and/or chunks |
Infrared spectrum | Conforms |
NMR | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...