cis-Dichlorobis(2,2'-bipyridine)ruthenium(II) dihydrate 99%

Supplier: Thermo Scientific Chemicals
Warning

AC36330-0010
TS36330-0010EA 208 USD
TS36330-0010
cis-Dichlorobis(2,2'-bipyridine)ruthenium(II) dihydrate 99%
cis-Dichlorobis(2,2'-bipyridine)ruthenium(II) dihydrate
Formula: C₂₀H₂₀Cl₂N₄O₂Ru
Storage Temperature: Ambient
MDL Number: MFCD00015504
CAS Number: 15746-57-3

Order Now

Specification Test Results

Appearance Black Powder or crystals
Infrared spectrum Conforms
Elemental Analysis ≥98.5 % (ex Ru)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR