N-Boc-amino-(4-N-boc-piperidinyl)carboxylic acid 95%

Warning

Synonyms: 1-Boc-4-(Boc-amino)piperidine-4-carboxylic Acid

36045-5000
TS36045-5000EA 491 USD
TS36045-5000
N-Boc-amino-(4-N-boc-piperidinyl)carboxylic acid 95%
N-Boc-amino-(4-N-boc-piperidinyl)carboxylic acid
Formula: C₁₆H₂₈N₂O₆
MW: 344.408 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02683116
CAS Number: 189321-65-1

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
HPLC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR