3-Bromophenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

35894-0010 35894-0050
TS35894-0010EA 32 USD
TS35894-0010 TS35894-0050
3-Bromophenylboronic acid 97%
3-Bromophenylboronic acid
Formula: C₆H₆BBrO₂
Melting Pt: 175…177 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00239386
CAS Number: 89598-96-9

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR