3-Bromophenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
35894-0010
35894-0050
TS35894-0010EA
32
USD
TS35894-0010
TS35894-0050
3-Bromophenylboronic acid 97%
3-Bromophenylboronic acid
Formula:
C₆H₆BBrO₂ Melting Pt: 175…177 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00239386 CAS Number: 89598-96-9 |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Titration with NaOH | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...