5,5'-Dimethyl-2,2'-bithiophene 99%

Supplier: Thermo Scientific Chemicals

35795-0010
TS35795-0010EA 74 USD
TS35795-0010
5,5'-Dimethyl-2,2'-bithiophene 99%
5,5'-Dimethyl-2,2'-bithiophene
Formula: C₁₀H₁₀S₂
Melting Pt: 65…68 °C
Storage Temperature: Refrigerator
CAS Number: 16303-58-5

Order Now

Specification Test Results

Appearance Yellow to light brown Crystals
Infrared spectrum Conforms
Melting point 65 to 68 °C
GC ≥99.9 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR