2-Bromo-3-methylpyridine 95%

Supplier: Thermo Scientific Chemicals
Warning

35766-0050
TS35766-0050EA 82 USD
TS35766-0050 TS35766-0250
2-Bromo-3-methylpyridine 95%
2-Bromo-3-methylpyridine
Formula: C₆H₆BrN
Flash Pt: >110 °C
MDL Number: MFCD00239380
CAS Number: 3430-17-9

Order Now

Specification Test Results

Appearance Clear light yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5650 to 1.5700 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR