1,1,1,3,3,3-Hexafluoro-2-propanol 99.8% for peptide synthesis

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,1,1,3,3,3-Hexafluoroisopropyl alcohol, Hexafluoro-2-propanol, Hexafluoroisopropanol, 1,1,1,3,3,3-Hexafluoropropan-2-ol, HFIP

35486-1000
TS35486-1000EA 1089 USD
TS35486-1000
1,1,1,3,3,3-Hexafluoro-2-propanol 99.8% for peptide synthesis
1,1,1,3,3,3-Hexafluoro-2-propanol
Formula: C₃H₂F₆O
MW: 168.04 g/mol
Boiling Pt: 59.1 °C (1013 hPa)
Melting Pt: –4 °C
Density: 1.596 g/cm³
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00011651
CAS Number: 920-66-1
EINECS: 213-059-4
UN: 3265
ADR: 8,I
Merck Index: 13,09598

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.02 % (K.F.)
GC ≥99.8 %
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %
Free amine ≤0.0002 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR