3,5-Dimethyl-4-hydroxybenzonitrile 97%

Warning

Synonyms: 4-Hydroxy-3,5-dimethylbenzonitrile

35366-0050
TS35366-0050EA 77 USD
TS35366-0050
3,5-Dimethyl-4-hydroxybenzonitrile 97%
3,5-Dimethyl-4-hydroxybenzonitrile
Formula: C₉H₉NO
MW: 147.18 g/mol
Melting Pt: 123…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00221716
CAS Number: 4198-90-7

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Melting point 123 to 126 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR