Methanol-[D₄] 100.0 atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals
Danger

351460075
TS351460075EA 272 USD
TS351460075
Methanol-[D₄] 100.0 atom % D for NMR spectroscopy
Methanol-[D4]

Packaged in 0.75 ml ampoules

Formula: D₃COD
Boiling Pt: 65.3 °C (1013 hPa)
Melting Pt: –99 °C
Density: 0.888 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00044637
CAS Number: 811-98-3
EINECS: 212-378-6
UN: 1230
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Water ≤0.02 % (Coulometric)
D-Enrichment ≥99.95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR