Deuterium oxide 99.8 atom % D for NMR spectroscopy

Supplier: Thermo Scientific Chemicals

351430075
TS351430075EA 37 USD
TS351430075
Deuterium oxide 99.8 atom % D for NMR spectroscopy
Deuterium oxide

Packaged in 0.75ml ampoules

Formula: D₂O
Boiling Pt: 101.3 °C (1013 hPa)
Melting Pt: 3.8 °C
Density: 1.105 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD00044636
CAS Number: 7789-20-0
EINECS: 232-148-9

Order Now

Specification Test Results

Appearance Clear colorless Liquid
D-Enrichment ≥99.75 %
Tritium (T) ≤370 Bq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR