Triethylammonium dihydrogentrifluoride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Triethylamine trihydrofluoride, Triethylamine trishydrofluoride

35062-0250 35062-1000
TS35062-0250EA 87 USD
TS35062-0250 TS35062-1000
Triethylammonium dihydrogentrifluoride 97%
Triethylammonium dihydrogentrifluoride
Formula: C₆H₁₈F₃N
MW: 161.21
Boiling Pt: 70 °C (15 mmHg)
Melting Pt: –29 °C
Density: 0.989 g/cm³
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00043294
CAS Number: 73602-61-6
EINECS: 277-550-5
UN: 2922
ADR: 8 I

Order Now

Specification Test Results

Appearance Colorless to yellow to orange Liquid
Infrared spectrum Conforms
Titration with NaOH 20 to 25 % (available HF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR