D-ɑ-Tocopherylquinone 90-95%

Supplier: Thermo Scientific Chemicals

Synonyms: Vitamin E Quinone (D-form)

35030-5000
TS35030-5000EA 449 USD
TS35030-5000
D-ɑ-Tocopherylquinone 90-95%
D-alpha-Tocopherylquinone
Formula: C₂₉H₅₀O₃
MW: 446.72 g/mol
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00135657
CAS Number: 7559-04-8

Order Now

Specification Test Results

Appearance Clear amber to red Viscous liquid
Infrared spectrum Conforms
GC ≥90 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR