Succinimide 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pyrrolidine-2,5-dione

34865-0050 34865-1000 34865-5000
TS34865-0050EA 38 USD
TS34865-0050 TS34865-1000 TS34865-5000
Succinimide 98%
Succinimide
Formula: C₄H₅NO₂
MW: 99.09 g/mol
Boiling Pt: 285
Melting Pt: 121…126 °C
Flash Pt: 201 °C
Storage Temperature: Ambient
MDL Number: MFCD00005495
CAS Number: 123-56-8
EINECS: 204-635-6
Merck Index: 12,09040

Order Now

Specification Test Results

Appearance Off-white to beige to light brown Powder or flakes
Infrared spectrum Conforms
Melting point 121 to 126 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR