4-Aminobenzyl alcohol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(Hydroxymethyl)aniline, (4-Aminophenyl)methanol
34824-0050
34824-0500
TS34824-0050EA
111
USD
TS34824-0050
TS34824-0500
4-Aminobenzyl alcohol 98%
4-Aminobenzyl alcohol
Formula:
C₇H₉NO MW: 123.15 Boiling Pt: 169…171 °C (11 mmHg) Melting Pt: 75 Storage Temperature: Refrigerator |
MDL Number:
MFCD00014782 CAS Number: 623-04-1 EINECS: 210-767-5 |
Specification Test Results
Appearance | Off-white to yellow to brown Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 59 to 65 °C |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...