4-Aminobenzyl alcohol 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Hydroxymethyl)aniline, (4-Aminophenyl)methanol

34824-0050 34824-0500
TS34824-0050EA 111 USD
TS34824-0050 TS34824-0500
4-Aminobenzyl alcohol 98%
4-Aminobenzyl alcohol
Formula: C₇H₉NO
MW: 123.15
Boiling Pt: 169…171 °C (11 mmHg)
Melting Pt: 75
Storage Temperature: Refrigerator
MDL Number: MFCD00014782
CAS Number: 623-04-1
EINECS: 210-767-5

Order Now

Specification Test Results

Appearance Off-white to yellow to brown Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 59 to 65 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR