Water, extra pure, deionized

Supplier: Thermo Scientific Chemicals

345470050 345470250
TS345470050EA 57 USD
TS345470050 TS345470250
Water, extra pure, deionized
Water
Formula: H₂O
Boiling Pt: 100 °C (760 mmHg)
Density: 1 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Residue after evaporation ≤25 ppm
Conductivity ≤5.0 µS/cm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR