(S)-(+)-1-Aminoindan 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (S)-(+)-1-Indanylamine

34253-0010
TS34253-0010EA 166 USD
TS34253-0010
(S)-(+)-1-Aminoindan 97%
(S)-(+)-1-Aminoindan
Formula: C₉H₁₁N
Boiling Pt: 96 °C (8 mmHg)
Melting Pt: 15 °C
Density: 1.038 g/cm³
Flash Pt: 94 °C
Storage Temperature: Ambient
MDL Number: MFCD00216670
CAS Number: 61341-86-4

Order Now

Specification Test Results

Appearance Clear yellow to light brown Liquid
Infrared spectrum Conforms
GC ≥96.0 %
Specific optical rotation +15° to +25° (20°C, 589 nm) (c=1.5, CH3OH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR