Sodium perchlorate 98%, Extra Pure
Supplier: Thermo Scientific Chemicals
|
Danger
|
34218-0025
AC34218-0010
AC34218-2500
TS34218-0025EA
514
USD
TS34218-0025
TS34218-0010
TS34218-2500
Sodium perchlorate 98%, Extra Pure
Sodium perchlorate
Formula:
ClNaO₄ Melting Pt: 482 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003480 CAS Number: 7601-89-0 EINECS: 231-511-9 UN: 1502 ADR: 5.1,II Merck Index: 13,08726 |
Specification Test Results
Appearance | White Powder or crystals and/or chunks |
pH | 6.0 to 8.0 (5% in water, 25°C) |
Titration after Ion exchange | 97.5 to 102.5 % |
Insoluble matter in water | ≤0.005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...