Sodium perchlorate 98%, Extra Pure

Supplier: Thermo Scientific Chemicals
Danger

34218-0025 AC34218-0010 AC34218-2500
TS34218-0025EA 514 USD
TS34218-0025 TS34218-0010 TS34218-2500
Sodium perchlorate 98%, Extra Pure
Sodium perchlorate
Formula: ClNaO₄
Melting Pt: 482 °C
Storage Temperature: Ambient
MDL Number: MFCD00003480
CAS Number: 7601-89-0
EINECS: 231-511-9
UN: 1502
ADR: 5.1,II
Merck Index: 13,08726

Order Now

Specification Test Results

Appearance White Powder or crystals and/or chunks
pH 6.0 to 8.0 (5% in water, 25°C)
Titration after Ion exchange 97.5 to 102.5 %
Insoluble matter in water ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR