Hexadecyl palmitate 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Cetyl Palmitate, Palmitic acid palmityl ester, n-Hexadecyl palmitate, Palmitic Acid Hexadecyl Ester, Palmityl palmitate
33846-0050
33846-0250
33846-1000
TS33846-0050EA
51
USD
TS33846-0050
TS33846-0250
TS33846-1000
Hexadecyl palmitate 98%
Hexadecyl palmitate
Formula:
C₃₂H₆₄O₂ MW: 480.86 g/mol Boiling Pt: 360 °C (760 mmHg) Melting Pt: 52…56 °C Storage Temperature: Ambient |
MDL Number:
MFCD00053739 CAS Number: 540-10-3 |
Specification Test Results
Appearance | White to almost white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 52 to 56 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...