D-erythro-Sphingosine (synthetic) 99+%

Supplier: Thermo Scientific Chemicals

Synonyms: D-Sphingosine, trans-D-erythro-2-Amino-4-octadecene-1,3-diol, (2S,3R,E)-2-Amino-4-octadecene-1,3-diol, 4-Sphingenine, (2S,3R,E)-2-Aminooctadec-4-ene-1,3-diol

33153-0500 33153-2500
TS33153-0500EA 388 USD
TS33153-0500 TS33153-2500
D-erythro-Sphingosine (synthetic) 99+%
D-erythro-Sphingosine
Formula: C₁₈H₃₇NO₂
MW: 299.50 g/mol
Melting Pt: 81…82 °C
Storage Temperature: Freezer
MDL Number: MFCD00036751
CAS Number: 123-78-4

Order Now

Specification Test Results

Appearance White to cream Powder or waxy solid
Infrared spectrum Conforms
TLC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR