N-Methyl-D-aspartic acid 98%

Supplier: Thermo Scientific Chemicals

32919-0500 32919-2500
TS32919-0500EA 82 USD
TS32919-0500 TS32919-2500
N-Methyl-D-aspartic acid 98%
N-Methyl-D-aspartic acid
Formula: C₅H₉NO₄
Melting Pt: 187…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00004226
CAS Number: 6384-92-5

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Specific optical rotation -26° to -30° (20°C, 589 nm) (c=1, 6 N HCl)
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR