DETA NONOate (NOC-18) 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: DETA/NO, 2,2′-(Hydroxynitrosohydrazino)bis-ethanamine, Diethylenetriamine NONOate, 3,3-Bis(aminoethyl)-1-hydroxy-2-oxo-1-triazene

32865-0250 32865-1000
TS32865-0250EA 99 USD
TS32865-0250 TS32865-1000
DETA NONOate (NOC-18) 95%
DETA NONOate (NOC-18)
Formula: C₄H₁₃N₅O₂
Melting Pt: 98…106 °C
Storage Temperature: Freezer
MDL Number: MFCD00278808
CAS Number: 146724-94-9

Order Now

Specification Test Results

Appearance White to off-white Crystalline solid
Infrared spectrum Conforms
NMR ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR