Suramin hexasodium 98%

Supplier: Thermo Scientific Chemicals

32854-0500
TS32854-0500EA 126 USD
TS32854-0500
Suramin hexasodium 98%
Suramin hexasodium
Formula: C₅₁H₃₄N₆Na₆O₂₃S₆
Storage Temperature: Refrigerator
MDL Number: MFCD00210217
CAS Number: 129-46-4

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR