Butyl acetate 99.5+% for electronic use (MOS) residue-free

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Butyl ethanoate, Acetic acid butyl ester, Acetic acid n-butyl ester, n-Butyl acetate

327850010
TS327850010EA 81 USD
TS327850010
Butyl acetate 99.5+% for electronic use (MOS) residue-free
Butyl acetate
Formula: CH₃COO(CH₂)₃CH₃
MW: 116.16 g/mol
Boiling Pt: 126 °C (1013 hPa)
Melting Pt: –78 °C
Density: 0.88 g/cm³
Flash Pt: 27 °C
Storage Temperature: Ambient
MDL Number: MFCD00009445
CAS Number: 123-86-4
EINECS: 204-658-1
UN: 1123
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.1 % (Coulometric)
GC ≥99.50 %
Color scale ≤15 APHA
Residue after evaporation ≤0.001 %
Arsenic (As) ≤0.01 ppm
Barium (Ba) ≤0.01 ppm
Copper (Cu) ≤0.02 ppm
Iron (Fe) ≤0.02 ppm
Heavy metals (as Pb) ≤50 ppm
Acidity (CH3COOH) ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR