Bicine 99+% for biochemistry

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(Bis(2-hydroxyethyl)amino)acetic acid, N,N-Bis(2-hydroxyethyl)glycine

32771-0250 32771-1000 32771-5000
TS32771-0250EA 44 USD
TS32771-0250 TS32771-1000 TS32771-5000
Bicine 99+% for biochemistry
Bicine
Formula: C₆H₁₃NO₄
Melting Pt: 190…192 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00004295
CAS Number: 150-25-4
EINECS: 205-755-1

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals or needles
Infrared spectrum Conforms
Water ≤1 % (K.F.)
Sulfated ash ≤0.1 %
UV at 280 nm A: ≤0.05
Titration with NaOH ≥99.0 %
Iron (Fe) ≤5 ppm
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR