N-(2-Acetamido)-2-aminoethanesulfonic acid (ACES) 99+% for biochemistry

Supplier: Thermo Scientific Chemicals

Synonyms: ACES, N-(Carbamoylmethyl)-2-aminoethanesulfonic Acid, N-(Carbamoylmethyl)taurine, N-(Carbamoylmethyl)-2-aminoethanesulfonic acid, N-(2-Acetamido)-2-aminoethanesulphonic acid, N-(Carbamoylmethyl)-2-aminoethanesulphonic Acid, 2-[(Carbamoylmethyl)amino]ethanesulfonic acid, N-(2-Acetamido)-2-aminoethanesulfonic acid

32767-0010 32767-1000 32767-2500
TS32767-0010EA 978 USD
TS32767-0010 TS32767-1000 TS32767-2500
N-(2-Acetamido)-2-aminoethanesulfonic acid (ACES) 99+% for biochemistry
N-(2-Acetamido)-2-aminoethanesulphonic acid (ACES)
Formula: H₂NCOCH₂NHCH₂CH₂SO₃H
Melting Pt: 270…295 °C
Storage Temperature: Ambient
MDL Number: MFCD00008030
CAS Number: 7365-82-4
EINECS: 230-908-4
Merck Index: 13,00037
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d8d301fc0de89d24309fbfdbaedb924c1debb4a&vid=a5fb2045198b642d124372f31d901db2e2d25597&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤1 % (K.F.)
Heavy metals ≤5 ppm
UV at 280 nm A: ≤0.02
pH 3.6 to 4.4 (1 % at 25°C)
Titration with NaOH ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR