γ-(N-Morpholino)propanesulfonic acid (MOPS) 99.5% for molecular biology, for peptide sequencing DNase-, RNase-, protease-free

Supplier: Thermo Scientific Chemicals

32766-5000 32766-1000
TS32766-5000EA 761 USD
TS32766-5000 TS32766-1000
γ-(N-Morpholino)propanesulfonic acid (MOPS) 99.5% for molecular biology, for peptide sequencing DNase-, RNase-, protease-free
γ-(N-Morpholino)propanesulphonic acid (MOPS)
Formula: C₇H₁₅NO₄S
Melting Pt: 277…282 °C
Flash Pt: 116 °C
Storage Temperature: Ambient
MDL Number: MFCD00006183
CAS Number: 1132-61-2
EINECS: 214-478-5
Merck Index: 13,06288

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Assay ≥99.5 %
Loss on drying ≤0.5 % (105°C)
Solubility (10 % in water) Complete
UV at 280 nm A: ≤0.01
pH 3.0 to 4.5 (1 % at 25°C)
Dnase,Rnase,Protease activity none detected
Heavy metals (as Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR