Tris Borate EDTA Buffer, for Molecular Biology, 10X, DNAse, RNAse and Protease free, pH 8.3, Thermo Scientific
Supplier: Thermo Scientific Chemicals
32733-0010
TS32733-0010EA
87
USD
TS32733-0010
Tris Borate EDTA Buffer, for Molecular Biology, 10X, DNAse, RNAse and Protease free, pH 8.3, Thermo Scientific
Buffers
Electrophoresis Buffers
Running Buffers
CAS No.: 610769-35-2
- MDL No.: MFCD00004679
- GHS Symbols: GHS07
- Appearance: Clear colorless Solution
- Storage Temperature: Ambient
- Signal word: Warning
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...