Sodium dodecyl sulfate (SDS) 99+% for molecular biology DNase-, RNase-, protease-free
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
32731-5000
32731-0025
32731-1000
TS32731-5000EA
470
USD
TS32731-5000
TS32731-0025
TS32731-1000
Sodium dodecyl sulfate (SDS) 99+% for molecular biology DNase-, RNase-, protease-free
Sodium dodecyl sulphate (SDS)
Formula:
CH₃(CH₂)₁₁OSO₃Na Melting Pt: 206 °C Flash Pt: >150 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036175 CAS Number: 151-21-3 EINECS: 205-788-1 UN: 1325 ADR: 4.1,III Merck Index: 14,08636 |
|
Specification Test Results
Appearance | White Powder |
Infrared spectrum | Conforms |
Assay | ≥99.0 % |
UV | at 280 nm A: ≤0.05 |
Dnase,Rnase,Protease activity | None detected |
Chloride (Cl) | ≤1000 ppm |
Phosphate (PO4) | ≤5 ppm |
Copper (Cu) | ≤5 ppm |
Lead (Pb) | ≤5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...