Sodium dodecyl sulfate (SDS) 99+% for molecular biology DNase-, RNase-, protease-free

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium lauryl sulfate, Dodecyl sulfate sodium salt, Lauryl sulfate sodium salt, Dodecyl sulfuric acid sodium salt, Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

32731-5000 32731-0025 32731-1000
TS32731-5000EA 470 USD
TS32731-5000 TS32731-0025 TS32731-1000
Sodium dodecyl sulfate (SDS) 99+% for molecular biology DNase-, RNase-, protease-free
Sodium dodecyl sulphate (SDS)
Formula: CH₃(CH₂)₁₁OSO₃Na
Melting Pt: 206 °C
Flash Pt: >150 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1,III
Merck Index: 14,08636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Assay ≥99.0 %
UV at 280 nm A: ≤0.05
Dnase,Rnase,Protease activity None detected
Chloride (Cl) ≤1000 ppm
Phosphate (PO4) ≤5 ppm
Copper (Cu) ≤5 ppm
Lead (Pb) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR