EDTA disodium salt dihydrate 99+% for molecular biology DNase-, RNase-, protease-free

Supplier: Thermo Scientific Chemicals
Warning

32720-0025 32720-1000 32720-5000
TS32720-0025EA 510 USD
TS32720-0025 TS32720-1000 TS32720-5000
EDTA disodium salt dihydrate 99+% for molecular biology DNase-, RNase-, protease-free
EDTA disodium salt dihydrate
Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
Melting Pt: 252 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6
EINECS: 205-358-3
Merck Index: 12,03556
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White Crystalline solid
Infrared spectrum Conforms
Titration Complexometric ≥99.0 %
UV (0.1 M in water)
pH 4 to 5 (5 % at 25°C)
Dnase,Rnase,Protease activity none detected
Iron (Fe) ≤100 ppm
Heavy metals (as Pb) ≤10 ppm
Nitrilotriacetic acid ≤0.1 %
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR